  • Search Results
  • Assessment of testicular corticosterone biosynthesis in adult male rats.
Assessment of testicular corticosterone biosynthesis in adult male rats.
PloS one (2015-02-24)
Naoyuki Maeda, Sachi Tahata, Takeshi Yagi, Emi Tanaka, Kanae Masu, Michiko Sato, Satoko Haeno, Takenori Onaga, Hiroshi Yokota
ABSTRACT
Corticosterone is synthesized in the adrenal glands and is circulated throughout the body to perform regulatory functions in various tissues. The testis is known to synthesize and secrete testosterone and other androgens. We developed an accurate method to measure steroid content using liquid chromatography-mass spectrometry analysis. In the present study, significant levels of the precursor compounds of testosterone and corticosterone synthesis could be detected in rat testis using this method. After adrenalectomy, corticosterone remained in the blood and testicular tissue at approximately 1% of the amount present in the control testis. When the excised testicular tissue was washed and incubated with NADH, NADPH and progesterone, not only testosterone and its precursors but also 11-deoxycorticosterone and corticosterone were produced; the levels of 11-deoxycorticosterone and corticosterone increased with incubation time. The production rate of 11-deoxycorticosterone from progesterone was estimated to be approximately 1/20 that of 17-hydroxyprogesterone, and the corticosterone level was approximately 1/10 that of testosterone. These ratios coincided with those in the testicular tissue of the adrenalectomized rats, indicating that corticosterone was synthesized in the testis and not in the blood. A primary finding of this study was that corticosterone and testosterone were synthesized in a 1/10-20 ratio in the testis. It is concluded that corticosterone, which has various functions, such as the regulation of glycolysis and mediating spermatogenesis, is produced locally in the testis and that this the local production is convenient and functional to respond to local needs.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
5-Pregnen-3β-ol-20-one, ≥98%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%