  • Search Results
  • Pharmacokinetic interaction profile of riociguat, a new soluble guanylate cyclase stimulator, in vitro.
Pharmacokinetic interaction profile of riociguat, a new soluble guanylate cyclase stimulator, in vitro.
Pulmonary pharmacology & therapeutics (2014-03-25)
Verena Rickert, Walter Emil Haefeli, Johanna Weiss
ABSTRACT
Riociguat is a new soluble guanylate cyclase stimulator under development for pulmonary arterial hypertension and chronic thromboembolic pulmonary hypertension. So far, the interaction potential of riociguat with other drugs is nearly unknown. Therefore, we assessed in vitro the potency of riociguat to inhibit important drug metabolising enzymes (cytochrome P450 (CYP) 3A4, CYP2C19, and CYP2D6) and drug transporters (P-glycoprotein (P-gp/ABCB1), breast cancer resistance protein (BCRP/ABCG2), and organic anion transporting polypeptides (OATP) 1B1 and 1B3). In addition we evaluated its substrate characteristics for P-gp, BCRP, and the multidrug resistance-associated protein 1 (MRP1/ABCC1). We also assessed riociguat's inducing properties on important drug metabolising enzymes and transporters and investigated its ability to activate the pregnane-X-receptor (PXR). Riociguat was identified as a weak to moderate inhibitor of P-gp (f2-value: 11.7 ± 4.8 μM), BCRP (IC50 = 46.2 ± 20.3 μM), OATP1B1 (IC50 = 34.1 ± 3.15 μM), OATP1B3 (IC50 = 50.3 ± 7.5 μM), CYP2D6 (IC50 = 12.4 ± 0.74 μM), and CYP2C19 (IC50 = 46.1 ± 7.14 μM). Furthermore, it induced mRNA expression of BCRP/ABCG2 (3-fold at 20 μM) and to a lesser extent of CYP3A4 (2.3-fold at 20 μM), UGT1A4, and ABCB11. The only weak inducing properties were confirmed by weak activation of PXR. Considering its systemic concentrations its interaction potential as a perpetrator drug seems to be low. In contrast, our data suggest that riociguat is a P-gp substrate and might therefore act as a victim drug when co-administered with strong P-gp inductors or inhibitors.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥97% (HPLC), crystalline
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Rifampicin, ≥97% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)