  • Search Results
  • The effect of statin on epithelial-mesenchymal transition in peritoneal mesothelial cells.
The effect of statin on epithelial-mesenchymal transition in peritoneal mesothelial cells.
PloS one (2014-10-03)
Tae Ik Chang, Hye-Young Kang, Kyung Sik Kim, Sun Ha Lee, Bo Young Nam, Jisun Paeng, Seonghun Kim, Jung Tak Park, Tae-Hyun Yoo, Shin-Wook Kang, Seung Hyeok Han
ABSTRACT
Statins have recently been highlighted for their pleiotropic actions distinct from cholesterol-lowering effects. Despite this interest, it is currently unknown whether statin therapy inhibits peritoneal dialysis (PD)-related epithelial-mesenchymal transition (EMT). In vitro, human peritoneal mesothelial cells (HPMCs) were exposed to 5.6 mM glucose (NG) or 100 mM glucose (HG) with or without simvastatin (1 µM). In vivo, PD catheters were inserted into 32 Sprague-Dawley rats, and saline (C, n = 16) or 4.25% peritoneal dialysis fluid (PDF) (PD, n = 16) was infused for 4 weeks. Eight rats from each group were treated with 5 mg/kg/day of simvastatin intraperitoneally. Changes in the protein expression of EMT markers such as E-cadherin, α-SMA, Snail, and fibronectin in HPMCs and the peritoneum were evaluated by Western blot analysis and immunofluorescence or immunohistochemical staining. We also explored whether activation of the mevalonate pathway and its downstream small GTPases were involved in dialysis-related peritoneal EMT and could be inhibited by statin treatment. Compared to NG cells, E-cadherin expression was significantly decreased, while α-SMA, Snail, and fibronectin expression were significantly increased in HPMCs exposed to HG, and these changes were abrogated by simvastatin (p<0.05). In addition, the cobblestone-like appearance of normal HPMCs was converted into a fibroblast-like morphology after HG treatment, which was reversed by simvastatin. These EMT-like changes were also observed in HPMCs treated with geranyl-geranyl pyrophosphate (5 µM). HG significantly increased the protein expression of RhoA and Rac1 in the membrane fractions, and these increases were ameliorated by simvastatin (p<0.05). In PD rats, E-cadherin in the peritoneum was significantly decreased, whereas α-SMA, Snail, and fibronectin expression were significantly increased (p<0.05) compared to C rats. The thickness of the mesothelial layer in the peritoneum were also significantly greater in PD rats than in C rats (p<0.05). These changes of the peritoneum in PD rats were significantly attenuated by simvastatin. This study demonstrated that PD-related EMT was mediated via the mevalonate pathway, and statin treatment inhibited the EMT changes in HG-treated HPMCs and PDF-stimulated PD rats. These findings suggest that statins may be a promising therapeutic strategy for preservation of peritoneal membrane integrity in long-term PD patients.
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Geranylgeranyl pyrophosphate ammonium salt, solution, ≥95% (TLC), ~1 mg/mL in methanol: NH4OH (7:3)
Supelco
Simvastatin, analytical standard
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Simvastatin, European Pharmacopoeia (EP) Reference Standard