  • Search Results
  • Decreased N(6)-methyladenosine in peripheral blood RNA from diabetic patients is associated with FTO expression rather than ALKBH5.
Decreased N(6)-methyladenosine in peripheral blood RNA from diabetic patients is associated with FTO expression rather than ALKBH5.
The Journal of clinical endocrinology and metabolism (2014-10-11)
Fan Shen, Wei Huang, Jing-Tao Huang, Jun Xiong, Ying Yang, Ke Wu, Gui-Fang Jia, Jinyun Chen, Yu-Qi Feng, Bi-Feng Yuan, Song-Mei Liu
ABSTRACT
N(6)-methyladenosine (m(6)A) modification plays a fundamental role in the epigenetic regulation of the mammalian transcriptome. m(6)A can be demethylated by fat mass- and obesity-associated (FTO) protein and α-ketoglutarate-dependent dioxygenase alkB homolog 5 (ALKBH5) protein. However, the importance of m(6)A alteration in type 2 diabetes mellitus (T2DM) has not been explored. The objective of the study was to investigate whether m(6)A content was reduced in T2DM patients and whether m(6)A content was correlated with the mRNA expression levels of the FTO and ALKBH5 genes. In this case-control study, peripheral blood samples were obtained from 88 T2DM patients and 92 healthy controls. For the diabetic animal model experiment, blood samples were obtained from seven diabetic and eight nondiabetic rats. A sensitive liquid chromatography-electrospray ionization-tandem mass spectrometry method was developed for the determination of the m(6)A content in RNA, quantitative real-time PCR was used to examine the mRNA expression levels of the FTO and ALKBH5 genes, and high-resolution melting and DNA sequencing were used to detect FTO single-nucleotide polymorphisms. Our results showed that the m(6)A contents in the RNA from T2DM patients and diabetic rats were significantly lower compared with the control groups (P = 2.6 × 10(-24) for T2DM patients; P = .001 for diabetic rats, respectively), and T2DM can be characterized by the content of m(6)A. The mRNA expression level of FTO was significantly higher in T2DM patients than that of the controls (P = .0007) and was associated with the risk of T2DM (odds ratio 2.797, 95% confidence interval 1.452-5.389, P = .002). Moreover, the m(6)A contents were correlated with FTO mRNA expression. These data suggest that the increased mRNA expression of FTO could be responsible for the reduction of m(6)A in T2DM, which may further increase the risk of complications of T2DM. Low m(6)A should be investigated further as a novel potential biomarker of T2DM.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%