  • Search Results
  • Functional crosstalk between the adenosine transporter CNT3 and purinergic receptors in the biliary epithelia.
Functional crosstalk between the adenosine transporter CNT3 and purinergic receptors in the biliary epithelia.
Journal of hepatology (2014-07-19)
Valeria Godoy, Jesús M Banales, Juan F Medina, Marçal Pastor-Anglada
ABSTRACT
Both hepatocytes and cholangiocytes release ATP into the bile, where it acts as a potent autocrine/paracrine stimulus that activates biliary secretory mechanisms. ATP is known to be metabolized into multiple breakdown products, ultimately yielding adenosine. However, the elements implicated in the adenosine-dependent purinergic regulation of cholangiocytes are not known. Normal rat cholangiocytes (NRCs) were used to study the expression of adenosine receptors and transporters and their functional interactions at the apical and basolateral membrane domains of polarized cholangiocytes. We found that: (1) cholangiocytes exclusively express two concentrative nucleoside transporters (CNT) known to be efficient adenosine carriers: CNT3, located at the apical membrane, and CNT2, located at apical and basolateral membrane domains; (2) in both domains, NRCs also express the high affinity adenosine receptor A2A, which modulated the activity of apical CNT3 in a domain-specific manner; (3) the regulation exerted by A2A on CNT3 was dependent upon the cAMP/PKA/ERK/CREB axis, intracellular trafficking mechanisms and AMPK phosphorylation; (4) secretin increased the activity of the apically-located CNT3, and promoted additional basolateral CNT3-related activity; and (5) extracellular ATP (a precursor of adenosine) was able to exert an inhibitory effect on the apical activity of both CNT3 and CNT2. This study uncovered the functional expression of nucleoside transporters in cholangiocytes and provides evidence for direct crosstalks between adenosine transporters and receptors for adenosine and its natural extracellular precursor, ATP. Our data anticipate the possibility of adenosine playing a major role in the physiopathology of the biliary epithelia.
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Cytidine, 99%
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Guanosine, ≥98%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications