  • Search Results
  • Cholinergic receptor activation on epithelia protects against cytokine-induced barrier dysfunction.
Cholinergic receptor activation on epithelia protects against cytokine-induced barrier dysfunction.
Acta physiologica (Oxford, England) (2015-02-17)
S Dhawan, I H Hiemstra, C Verseijden, F W Hilbers, A A Te Velde, L E M Willemsen, J Stap, J M den Haan, W J de Jonge
ABSTRACT
Various types of cholinergic receptors are expressed on intestinal epithelia. Their function is not completely understood. We hypothesize that cholinergic receptor activation on epithelium may serve a protective function in cytokine-induced barrier dysfunction. The effect of cholinergic receptor activation on cellular barrier function in epithelial cells was assessed by measuring electrical impedance, and by determining para-cellular transport in transwell experiments. Cell lysates treated with cytokine and/or cholinergic agonists were analysed for cyto- and chemokine production, and tight junction (TJ) protein rearrangement was assessed. Primary colonic epithelial cells were isolated from surgically resected colon tissue of patients with inflammatory bowel disease. IL-1β induced production of chemokines (CXCL-1, CXCL-10, IL-8, CCL-7) and led to a rearrangement of TJ proteins (occludin and ZO-1). This response was inhibited by pre-treatment with muscarinic, rather than nicotinic, acetylcholine receptor agonists. Treatment with IL-1β enhanced paracellular permeability (4kD dextran) and reduced impedance across the monolayer, which was counteracted by pre-incubation with acetylcholine, or muscarinic receptor agonist bethanechol. The protective effect of acetylcholine was antagonized by atropine, underscoring muscarinic receptor involvement. IL-1β induced transcription of myosin light chain kinase and phosphorylation of myosin light chain, and this cytokine-induced phosphorylation of MLC was inhibited by muscarinic receptor agonists. Furthermore, in epithelial cells from resection material of patients with Crohn's disease and ulcerative colitis, high expression of CXCL-8 was associated with a reduced choline acetyl transferase expression, suggesting an aberrant epithelial production of ACh in inflammatory context. Acetylcholine acts on muscarinic receptors on epithelial cells to maintain epithelial barrier function under inflammatory conditions.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Mineral oil, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)