MilliporeSigma
  • Drug use, sexual risk behaviour and sexually transmitted infections among swingers: a cross-sectional study in The Netherlands.
Drug use, sexual risk behaviour and sexually transmitted infections among swingers: a cross-sectional study in The Netherlands.
Sexually transmitted infections (2014-10-25)
Laura W L Spauwen, Anne-Marie Niekamp, Christian J P A Hoebe, Nicole H T M Dukers-Muijrers
ABSTRACT
Recreational drug use has been found to be associated with high-risk sexual behaviour and with sexually transmitted infections (STI). This study is the first to assess the prevalence of drug use among swingers (heterosexuals who, as a couple, practise mate swapping or group sex, and/or visit sex clubs for couples), and its association with high-risk sexual behaviour and STI. We recruited individuals who self-identified as swingers and visited our STI clinic (from 2009 to 2012, South Limburg, The Netherlands). Participants (n=289; median age 45 years; 49% female) filled in a self-administered questionnaire on their sexual and drug use behaviour while swinging, over the preceding 6 months. We assessed associations between sexual behaviour, drug use and STI diagnoses (Chlamydia trachomatis (CT), Neisseria gonorrhoeae (NG), syphilis, HIV and hepatitis B) using logistic regression analyses. Overall, the prevalence of CT and/or NG was 13%. No other STIs were observed. Seventy-nine percent of swingers reported recreational drug use (including alcohol and use of erectile dysfunction drugs); 46% of them reported multiple drug use. Recreational drug use excluding alcohol and erectile dysfunction drugs (reported by 48%) was associated with high-risk behaviours in men and women. Drug use was independently associated with STI in female swingers, especially those who practice group sex. High rates of multiple drug use, as well as risky sexual behaviour and STIs among swingers, warrant paying more attention to this key population in prevention and care, as they are a risk group that is generally under-recognised and underserved in care.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%