  • Search Results
  • Large-scale high-quality 2D silica crystals: dip-drawing formation and decoration with gold nanorods and nanospheres for SERS analysis.
Large-scale high-quality 2D silica crystals: dip-drawing formation and decoration with gold nanorods and nanospheres for SERS analysis.
Nanotechnology (2014-09-13)
Vitaly A Khanadeev, Boris N Khlebtsov, Svetlana A Klimova, Mikhail Yu Tsvetkov, Victor N Bagratashvili, Gleb B Sukhorukov, Nikolai G Khlebtsov
ABSTRACT
High-quality colloidal crystals (CCs) are important for use in photonic research and as templates for large-scale plasmonic SERS substrates. We investigated how variations in temperature, colloid concentration, and dip-drawing parameters (rate, incubation time, etc) affect the structure of 2D CCs formed by highly monodisperse silica nanoparticles (SiNPs) synthesized in an l-arginine solution and regrown by a modified Stöber method. The best quality 2D CCs were obtained with aqueous 12 wt% colloids at a temperature of 25 °C, an incubation time of 1 min, and a drawing rate of 50 mm min(-1). Assembling of gold nanorods (GNRs) on 2D CCs resulted in the formation of ring-like chains with a preferential tail-to-tail orientation along the hexagonal boundaries. To the best of our knowledge, this is the first time that such nanostructures have been prepared. Owing to the preferential tail-to-tail packing of GNRs, 2D SiNP CC + GNR substrates demonstrated an analytical SERS enhancement of about 8000, which was 10 to 15 times higher than that for self-assembled GNRs on a silicon wafer. In addition, the analytical SERS enhancement was almost 60 times lower after replacing the nanorods in 2D SiNP CC + GNR substrates with 25 nm gold nanospheres.
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
SAFC
Hexadecyltrimethylammonium bromide
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.