  • Search Results
  • A new analytical method to determine non-steroidal anti-inflammatory drugs in surface water using in situ derivatization combined with ultrasound-assisted emulsification microextraction followed by gas chromatography-mass spectrometry.
A new analytical method to determine non-steroidal anti-inflammatory drugs in surface water using in situ derivatization combined with ultrasound-assisted emulsification microextraction followed by gas chromatography-mass spectrometry.
Talanta (2014-08-17)
Cheong Hoon Lee, Yujin Shin, Min Woo Nam, Kyung Min Jeong, Jeongmi Lee
ABSTRACT
Because of the high stability and potential toxic effects of non-steroidal anti-inflammatory drugs (NSAIDs), it is important to closely monitor their concentrations in the environment using a sensitive analytical method. In this study, a simple, rapid, efficient, and sensitive analytical method based on gas chromatography-mass spectrometry (GC-MS) was developed to determine the levels of seven common NSAIDs in various types of surface water. To simplify sample preparation, in situ derivatization using methyl chloroformate was combined with ultrasound-assisted emulsification microextraction. For selection and optimization of significant variables, experiments were statistically designed using Plackett-Burman design and central composite design. The resulting optimal conditions for derivatization and extraction were 100 μL of chloroform (extraction solvent), 10.0 mL of sample, and 240 μL of pyridine (catalyst as a base in derivatization). The optimized sample preparation coupled with optimized GC-MS analysis in selected ion monitoring mode provided good linearity from 0.010 to 5.0 ng mL(-1), and a limit of detection between 0.0050 and 0.010 ng mL(-1), good intra-day and inter-day precision (0.30-6.3% and 5.1-9.5%, respectively), and good accuracy (relative recovery; 91-117% at 0.20 ng mL(-1) and 77-105% at 2.5 ng mL(-1)). Compared with previously reported methods, the current method requires a small volume of sample and simple sample preparation steps for sensitive determination of NSAID levels using a conventional GC-MS system. The method was successfully applied to determine the levels of seven common NSAIDs in various types of surface water.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl chloroformate, 97%
Sigma-Aldrich
Isobutyl chloroformate, 98%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.