  • Search Results
  • Perfluorinated compounds in serum and urine samples from children aged 5-13 years in South Korea.
Perfluorinated compounds in serum and urine samples from children aged 5-13 years in South Korea.
Environmental pollution (Barking, Essex : 1987) (2014-06-24)
Da-Hye Kim, Mi-Young Lee, Jeong-Eun Oh
ABSTRACT
Serum and urine samples from 120 children aged 5-13 years from Dae-gu, Korea, were analyzed for 16 perfluorinated compounds (PFCs). The total PFC concentrations in the serum were 4.26-29.70 ng/mL, and perfluorohexanesulfonate (PFHxS), perfluorooctanoic acid (PFOA), perfluorooctanesulfonate (PFOS, which was dominant overall, at 6.58 ng/mL), and perfluoroundecanoic acid (PFUndA) were detected in all serum samples. The total PFC concentrations in the urine ranged from below the detection limit to 14.9 ng/mL, and perfluoropentanoic acid (PFPeA) was predominant. The PFOS (p < 0.005) concentration was higher in the serum of children than that of Korean adults aged 20-29. Some of the PFC concentrations in the serum correlated negatively with body mass index and tended to increase with the duration of breastfeeding. However, there were no gender-specific differences in the PFC concentrations and no correlations between PFC concentrations in serum and urine.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%