  • Search Results
  • Determining in situ periphyton community responses to nutrient and atrazine gradients via pigment analysis.
Determining in situ periphyton community responses to nutrient and atrazine gradients via pigment analysis.
The Science of the total environment (2015-02-24)
Rebecca L Dalton, Céline Boutin, Frances R Pick
ABSTRACT
Agrochemicals, including fertilizers and herbicides, are significant contributors of non-point source pollution to surface waters and have the potential to negatively affect periphyton. We characterized periphyton communities using pigment markers to assess the effects of nutrient enrichment and the herbicide atrazine with in situ experimental manipulations and by examining changes in community structure along existing agrochemical gradients. In 2008, the addition of nutrients (20 mg/L nitrate and 1.25 mg/L reactive phosphate), atrazine (20 μg/L) and a combination of both nutrients and atrazine had no significant effect on periphyton biomass or community structure in a stream periphytometer experiment. In 2009, similar experiments with higher concentrations of atrazine (200 μg/L) at two stream sites led to some minor effects. In contrast, at the watershed scale (2010) periphyton biomass (mg/m(2) chlorophyll a) increased significantly along correlated gradients of nitrate and atrazine but no direct effects of reactive phosphate were observed. Across the watershed, the average periphyton community was composed of Bacillariophyceae (60.9%), Chlorophyceae (28.1%), Cryptophyceae (6.9%) and Euglenophyceae (4.1%), with the Bacillariophyceae associated with high turbidity and the Chlorophyceae with nitrate enrichment. Overall, effects of nitrate on periphyton biomass and community structure superseded effects of reactive phosphate and atrazine.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%