MilliporeSigma
Cerebrosides from a Far-Eastern Glass Sponge Aulosaccus sp.
Lipids (2014-12-17)
Еlena А Santalova, Vladimir А Denisenko, Pavel S Dmitrenok, Аnatoly L Drozdov, Valentin A Stonik
ABSTRACT
Nine new cerebrosides 1a-d, 2a, 2b, 3a-c were found in the extract of a Far-Eastern glass sponge Aulosaccus sp. (class Hexactinellida). These β-D-glucopyranosyl-(1 → 1)-ceramides contain sphingoid bases (2S,3S,4R,11Z)-2-aminoeicos-11-ene-1,3,4-triol (in 1a-d), (2S,3S,4R,13Z)-2-aminoeicos-13-ene-1,3,4-triol (in 2a, b) and (2S,3S,4R,13S*,14R*)-2-amino-13,14-methylene-eicosane-1,3,4-triol (in 3a-c), which are N-acylated by (2R,15Z)-2-hydroxydocos-15-enoic (in 1a, 2a, 3a), (2R,16Z)-2-hydroxytricos-16-enoic (in 1b, 2b, 3b), (2R,17Z)-2-hydroxytetracos-17-enoic (in 1d) and (2R)-2-hydroxydocosanoic (in 1c, 3c) acids. The monoenoic and cyclopropane-containing sphingoid bases of compounds 1a-d, 2a, 2b, 3a-c have not been found previously in any sphingolipids. The structures of the cerebrosides were elucidated on the basis of (1)H-, (13)C-NMR spectroscopy, mass spectrometry, optical rotation data and chemical transformations. A simplified method for the assignment of the absolute configuration of 2-hydroxy fatty acids by GC analysis of their (2R)- and (2S)-oct-2-yl esters was proposed.
Product Description
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology