  • Search Results
  • Essential oil from leaves of Liquidambar formosana ameliorates inflammatory response in lipopolysaccharide-activated mouse macrophages.
Essential oil from leaves of Liquidambar formosana ameliorates inflammatory response in lipopolysaccharide-activated mouse macrophages.
Natural product communications (2014-08-15)
Kuo-Feng Hua, Tzu-Jung Yang, Huan-Wen Chiu, Chen-Lung Ho
ABSTRACT
The essential oil from Liquidambar formosana leaves (EOLF) was demonstrated to exhibit anti-inflammatory activity in mouse macrophages. EOLF reduced nitrite oxide generation, secretion levels of tumor necrosis factor-alpha and interleukin-6, and expression levels of prointerleukin-beta, inducible nitric oxide synthase, and cyclooxygenase-2 in lipopolysaccharide (LPS)-activated mouse macrophages. EOLF also reduced NLRP3 inflammasome-derived interleukin-1beta secretion. The underlying mechanisms for the EOLF-mediated anti-inflammatory activity were (1) reduction of LPS-induced reactive oxygen species generation; (2) reduction of LPS-induced activation of c-Jun N-terminal kinase, extracellular signal-regulated kinase, and p38 MAP kinase; (3) reduction of LPS-induced nuclear factor-kappaBeta activation. Furthermore, 25 compounds were identified in the EOLF using GC-FID and GC-MS and the major compounds were terpinen-4-ol (32.0%), beta-pinene (18.0%), gamma-terpinene (13.8%), and alpha-terpinene (9.7%). We found that LPS-induced nitrite oxide generation was inhibited significantly by terpinen-4-ol. Our results indicated that EOLF has anti-inflammatory activity and may provide a molecular rationale for future therapeutic interventions in immune modulation.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Human IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant