  • Search Results
  • Transcription of the activating receptor NKG2D in natural killer cells is regulated by STAT3 tyrosine phosphorylation.
Transcription of the activating receptor NKG2D in natural killer cells is regulated by STAT3 tyrosine phosphorylation.
Blood (2014-06-04)
Shiguo Zhu, Prasad V Phatarpekar, Cecele J Denman, Vladimir V Senyukov, Srinivas S Somanchi, Hoainam T Nguyen-Jackson, Emily M Mace, Alexandra F Freeman, Stephanie S Watowich, Jordan S Orange, Steven M Holland, Dean A Lee
ABSTRACT
Signal transducer and activator of transcription 3 (STAT3) is considered a negative regulator of inflammation, as inhibition of STAT3 signaling enhances antitumor immunity. However, STAT3 activation is a key oncogenic pathway in natural killer (NK)-lineage large granular lymphomas, and we recently reported enhanced proliferation and function of human NK cells activated with IL-21, which signals primarily through STAT3. These IL-21-expanded NK cells also have increased NKG2D expression, which led us to focus our investigation on whether STAT3 regulates NKG2D. In this study, we show that modulation of STAT3 phosphorylation with cytokines and small-molecule inhibitors correlates with NKG2D expression on human NK cells, leading to altered NK-cell degranulation. Moreover, NKG2D expression on murine NK cells having conditional STAT3 ablation is lower than on NK cells from wild-type mice, and human NK cells carrying dominant-negative STAT3 mutations have decreased baseline NKG2D expression and blunted responses to IL-10 and IL-21. Lastly, we show binding of STAT3 to a predicted STAT3 binding site upstream of the NKG2D gene, which is enhanced by IL-10 and IL-21 and decreased by STAT3 inhibition. Taken together, these data show that NKG2D expression in NK cells is regulated at the transcriptional level by STAT3, resulting in a functional NK cell defect in patients with STAT3 mutations.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Cucurbitacin I hydrate, ≥95% (HPLC), solid
Sigma-Aldrich
S3I-201, ≥97% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
IL-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)