  • Search Results
  • Immunoproteomic characterisation of Mycoplasma mycoides subspecies capri by mass spectrometry analysis of two-dimensional electrophoresis spots and western blot.
Immunoproteomic characterisation of Mycoplasma mycoides subspecies capri by mass spectrometry analysis of two-dimensional electrophoresis spots and western blot.
The Journal of pharmacy and pharmacology (2014-12-17)
Colin P Churchward, Rubén S Rosales, Adriana Gielbert, Mercedes Domínguez, Robin A J Nicholas, Roger D Ayling
ABSTRACT
Mycoplasma mycoides subspecies capri is one of the causative agents of contagious agalactia in goats. The disease is characterised by mastitis, pneumonia, arthritis, keratitis and in acute cases septicaemia. No vaccine is currently available that has been demonstrated to prevent disease. This study used two-dimensional electrophoresis to separate proteins from whole-cell preparations and tandem mass spectrometry to identify them. In total, 145 spots were successfully identified corresponding to 74 protein identities. Twenty of these proteins were found to be immunogenic by western blot analysis using a pooled serum sample from experimentally infected goats. Six proteins were found to have a less than 95% amino acid similarity to a closely related Mycoplasma species showing that they warrant further evaluation in development of diagnostic tests. These proteins were a dihydrolipoamide acetyltransferase component of the pyruvate dehydrogenase complex, phosphoglycerate kinase, pyrimidine-nucleoside phosphorylase, 30S ribosomal protein S6, ribulose-phosphate 3-epimerase and D-lactate dehydrogenase.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%