  • Search Results
  • Mirtoselect, an anthocyanin-rich bilberry extract, attenuates non-alcoholic steatohepatitis and associated fibrosis in ApoE(∗)3Leiden mice.
Mirtoselect, an anthocyanin-rich bilberry extract, attenuates non-alcoholic steatohepatitis and associated fibrosis in ApoE(∗)3Leiden mice.
Journal of hepatology (2014-12-17)
Martine C Morrison, Wen Liang, Petra Mulder, Lars Verschuren, Elsbet Pieterman, Karin Toet, Peter Heeringa, Peter Y Wielinga, Teake Kooistra, Robert Kleemann
ABSTRACT
Anthocyanins may have beneficial effects on lipid metabolism and inflammation and are demonstrated to have hepatoprotective properties in models of restraint-stress- and chemically-induced liver damage. However, their potential to protect against non-alcoholic steatohepatitis (NASH) under conditions relevant for human pathogenesis remains unclear. Therefore, we studied the effects of the standardised anthocyanin-rich extract Mirtoselect on diet-induced NASH in a translational model of disease. ApoE(∗)3Leiden mice were fed a Western-type cholesterol-containing diet without (HC) or with 0.1% (w/w) Mirtoselect (HCM) for 20weeks to study the effects on diet-induced NASH. Mirtoselect attenuated HC-induced hepatic steatosis, as observed by decreased macro- and microvesicular hepatocellular lipid accumulation and reduced hepatic cholesteryl ester content. This anti-steatotic effect was accompanied by local anti-inflammatory effects in liver, as demonstrated by reduced inflammatory cell clusters and reduced neutrophil infiltration in HCM. On a molecular level, HC diet significantly induced hepatic expression of pro-inflammatory genes Tnf, Emr1, Ccl2, Mpo, Cxcl1, and Cxcl2 while this induction was less pronounced or significantly decreased in HCM. A similar quenching effect was observed for HC-induced pro-fibrotic genes, Acta2 and Col1a1 and this anti-fibrotic effect of Mirtoselect was confirmed histologically. Many of the pro-inflammatory and pro-fibrotic parameters positively correlated with intrahepatic free cholesterol levels. Mirtoselect significantly reduced accumulation and crystallisation of intrahepatic free cholesterol, providing a possible mechanism for the observed hepatoprotective effects. Mirtoselect attenuates development of NASH, reducing hepatic lipid accumulation, inflammation and fibrosis, possibly mediated by local anti-inflammatory effects associated with reduced accumulation and crystallisation of intrahepatic free cholesterol.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
SAFC
Cholesterol, Plant-Derived
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Sucrose, analytical standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, meets USP testing specifications
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications