  • Search Results
  • A preliminary risk assessment of potential exposure to naturally occurring estrogens from Beijing (China) market milk products.
A preliminary risk assessment of potential exposure to naturally occurring estrogens from Beijing (China) market milk products.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-06-10)
Chen Chen, Xiaoxia Mi, Yuwei Yuan, Gang Chen, Li Ren, Kaiqiang Wang, Dan Zhu, Yongzhong Qian
ABSTRACT
This study was conducted to determine the occurrence of the natural steroid hormones estrone (E1), 17α-estradiol (αE2), 17β-estradiol (βE2) and estriol (E3) in 38 commercial milk samples obtained from markets in Beijing, China. Liquid Chromatography coupled with tandem mass spectrometry (LC-MS/MS) was employed to determine estrogens levels. The concentrations of E1, αE2, βE2 and E3 in different milk products varied from 0-146.12 ng/L, 0-70.12 ng/L, 0-31.85 ng/L to 0-2.18 ng/L, respectively. We compared exposures to estrogens through milk consumption with acceptable daily intakes (ADIs) and threshold for toxicological concern (TTC) to determine whether estrogen intakes from milk consumption are larger or smaller than the toxicity-based benchmarks. The combined margin of safety MOS (MOST) for total estrogens are about 72-99, 118-161, 539-1104, for 2-4, 4-7 year-old residential children, and adults, respectively. The lowest MOST for children of 2-4 years old result from comparing total of estrogens with the lowest TTC value (0.15 μg/person/day) (MOS=3.5). The MOS values suggest that the individual and total estrogens that may present in milk are not causing a health risk for the local residents, including young children.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dansyl chloride, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles