  • Search Results
  • Determination of pharmaceuticals in bivalves using QuEChERS extraction and liquid chromatography-tandem mass spectrometry.
Determination of pharmaceuticals in bivalves using QuEChERS extraction and liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2015-03-18)
Mireia Núñez, Francesc Borrull, Núria Fontanals, Eva Pocurull
ABSTRACT
A method for the quantitative determination of seven pharmaceuticals in bivalves was developed by QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) extraction, followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS) with electrospray ionization. Both the European Standard Method EN 15662 and the AOAC Official Method 2007.01 for QuEChERS were tested. In addition, several clean-up strategies were evaluated in order to clean the matrix previous to the LC-MS/MS analyses. Dispersive solid-phase extraction with silica gel and modification of the chromatographic separation were the clean-up strategies that gave the best results. The optimized method was validated in mussels (Mytilus galloprovincialis) and allowed the determination of pharmaceuticals at nanograms per gram levels (dry weight (d.w.)). Limits of quantification ranged from 5 to 100 ng/g. Apparent recoveries ranged from 35 to 77%. The application of this method to bivalves revealed the presence of salicylic acid at concentrations up to 103 ng/g (d.w.).
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis