  • Search Results
  • Anti-inflammatory activity of methanol extract and n-hexane fraction mojabanchromanol b from Myagropsis myagroides.
Anti-inflammatory activity of methanol extract and n-hexane fraction mojabanchromanol b from Myagropsis myagroides.
Life sciences (2014-08-15)
Da-Hyun Jeong, Koth-Bong-Woo-Ri Kim, Min-Ji Kim, Bo-Kyeong Kang, Dong-Hyun Ahn
ABSTRACT
This study was carried out to verify the anti-inflammatory effect of methanol extract from Myagropsis myagroides (MMME) and its n-hexane fraction mojabanchromanol b. The murine macrophages Raw264.7 cells were used. The pro-inflammatory cytokines (IL-6, IL-1β, TNF-α) and the expression of iNOS, COX-2, and NF-κB p65 were examined by ELISA and immunoblotting. To investigate the inhibitory effect of MMME in an animal model of inflammation, an assay to determine croton oil-induced ear edema in mice was performed. NO levels decreased with increasing concentration of MMME, and were inhibited up to 50%. The secretion of IL-6, TNF-α, and IL-1β was suppressed in a dose-dependent manner, especially at 50μg/mL, inhibition activities of cytokines were over 50%. MMME also suppressed the expression of COX-2, iNOS, and NF-κB p65, suggesting that MMME could affect the expression of inflammation related cytokines and proteins through the deregulation of NF-κB. Moreover, the formation of mouse ear edema was reduced at the highest dose tested compared to that in the control, and generated similar effects compared with prednisolone at 250mg/kg in mice ear edema evaluation test. In addition, the results in photomicrograph of mice ear tissue and mast cells also showed the same effect. After purification of fractions of MMME, it indicated that n-hexane fraction mojabanchromanol b was the most active fraction showing the inhibitory effect of IL-6 and TNF-α. These results suggested that MMME and mojabanchromanol b may have great effects on inflammatory factors and be potential anti-inflammatory therapeutic materials.
Sigma-Aldrich
Anti-RELA (Ab-311) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-529) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-536) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-435) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-254) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-276) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-505) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Anti-RELA (Ab-468) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard