Anti-inflammatory activity of lanoconazole, a topical antifungal agent.
Mycoses (2015-02-14)
Hideya Uratsuji, Aki Nakamura, Yoshihito Yamada, Kei Hashimoto, Tatsumi Matsumoto, Fumiaki Ikeda, Ritsuko Ishii
ABSTRACT
Topical antifungal agents which have anti-inflammatory effects have the potential to provide additional clinical benefits. Therefore, an anti-inflammatory activity of lanoconazole (LCZ), a topical antifungal agent, was investigated against in vitro and in vivo models of inflammation. The release of interleukin-8 (IL-8) from human epidermal keratinocytes stimulated by the addition of 100 μg ml(-1) β-glucan of Saccharomyces cerevisiae was significantly inhibited by LCZ at the concentration of 10(-5) mol l(-1). The release of interferon-γ and IL-2 from human peripheral blood mononuclear cells stimulated by the addition of 30 and 100 μg ml(-1) phytohemagglutinin was significantly inhibited by LCZ at the concentrations of 10(-7) and 10(-6) mol l(-1), respectively. The increase in the ear thickness induced by topical application of 0.01% 12-O-tetradecanoyl phorbol-13-acetate and 1% 2,4,6-trinitrochlorobenzene (TNCB) after sensitisation with 3% TNCB were established as the mouse models of irritant and contact dermatitis, respectively. Application of 1% and 3% LCZ showed a significant anti-inflammatory activity against both the irritant and contact dermatitis models. These findings suggest that LCZ possesses an anti-inflammatory activity, which may be partially helpful in the treatment of dermatomycoses.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Tris(2-pyridylmethyl)amine, 98%
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Betamethasone, ≥98%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Supelco
Betamethasone, VETRANAL®, analytical standard
Betamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Betamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Betamethasone, meets USP testing specifications