  • Search Results
  • Long-term sevelamer treatment lowers serum fibroblast growth factor 23 accompanied with increasing serum Klotho levels in chronic haemodialysis patients.
Long-term sevelamer treatment lowers serum fibroblast growth factor 23 accompanied with increasing serum Klotho levels in chronic haemodialysis patients.
Nephrology (Carlton, Vic.) (2014-08-13)
Hsin-Hung Lin, Hung-Hsiang Liou, Ming-Shiou Wu, Ching-Yuang Lin, Chiu-Ching Huang
ABSTRACT
Fibroblast growth factor 23 (FGF23) and Klotho are associated with vascular calcification and cardiovascular disease in dialysis patients. Sevelamer has been shown to reduce progression of vascular calcification. This study aimed to determine the long-term effect of sevelamer treatment on serum FGF23 and Klotho levels in chronic haemodialysis (HD) patients. In the post-hoc analysis, we measured serum FGF23, Klotho and other biochemical factors (Ca, P, i-PTH, hsCRP, LDL-C) in 50 haemodialysis patients, who completed a 48-week, open-Label, controlled randomized parallel-group study. Twenty-three patients received sevelamer and 27 patients received calcium carbonate. After 48-week sevelamer treatment, there were significant changes with lower LDL-C (from 2.82 ± 0.78 to 1.65 ± 0.53 mmol/L, P = 0.000), lower FGF23 (from 2465.97 (2568.88) to 795.61 (1098.39), P = 0.000) and higher s-Klotho levels (from 189.35 (161.88) to 252.94 (517.80) pg/mL, P = 0.000). In calcium carbonate group, there were no significant changes of LDL-C and FGF23, but with a borderline significant increase of s-Klotho level (from 142.34 (265.24) to 188.57 (252.38) pg/mL, P = 0.054). Multivariate analysis showed that FGF23 decrement was associated with sevelamer treatment (β = -0.277, P = 0.005), change of serum phosphate (β = 0.609, P = 0.000) and calcium levels (β = 0.635, P = 0.000). The increase of serum Klotho was associated with the decrease of serum phosphate (β = 0.490, P = 0.019). Maintenance HD patients had lower serum FGF23 levels, accompanied with significantly increased serum Klotho levels, after 48-week sevelamer treatment. The FGF23 decrement was associated with sevelamer use, the change of serum phosphate and calcium levels. The serum Klotho increment was proportional to the phosphate-lowering power of the binders.
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%