The effect of cyclosporine A in hemorrhagic shock model of rats.
The journal of trauma and acute care surgery (2015-03-11)
Kyuseok Kim, Jung Ho Shin, Jae Hyuk Lee, You Hwan Jo, Min A Kim, Kyoungbun Lee, Kyeong Won Kang, Yun-Sik Hong
ABSTRACT
The inhibition of mitochondrial permeability transition pore opening during ischemia-reperfusion can ameliorate injuries. This study aimed to investigate the effects of cyclosporine A (CsA) in rats after hemorrhagic shock. Male Sprague-Dawley rats were subjected to pressure-controlled hemorrhagic shock (mean arterial pressure, 38 ± 1 mm Hg) for 90 minutes. After the hemorrhagic shock period, rats were randomly allocated to one of three groups as follows: a control group, a CsA10 group, or a CsA50 group. CsA for the treatment groups (10 mg/kg for the CsA10 group and 50 mg/kg for the CsA50 group) or normal saline for the control group was administered via tail vein for 10 minutes, and shed blood was transfused for 15 minutes. For the survival study, animals were observed for up to 9 hours, and their survival time was recorded until death. Separate experiments were performed to examine the effect of CsA on inflammatory responses and liver injury. Rats were sacrificed at 210 minutes after the shock period, and blood and liver tissues were harvested. Survival times were shown to be significantly longer in the CsA-treated groups (i.e., the CsA10 and CsA50 groups) than in the control group. Plasma interleukin-6 and thiobarbituric acid-reactive substances were significantly lower in the CsA50 group than in the control group and phosphorylation of Akt, GSK-3β, and Bad were significantly increased in the CsA-treated groups compared with the control group. Expressions of Bcl-2, cleaved caspase 3, and cytoplasmic cytochrome C were significantly decreased in the CsA-treated groups compared with the control group. Although histologic liver injury was not significantly different among the groups, ultrastructural changes of mitochondria were more prominent in the control group than in the CsA-treated groups. CsA increased survival time, decreased proinflammatory cytokine and lipid peroxidation, and augmented Akt survival pathways in rats subjected to pressure-controlled hemorrhagic shock.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Xylazine, ≥99%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri