  • Search Results
  • The phosphate clamp: sequence selective nucleic acid binding profiles and conformational induction of endonuclease inhibition by cationic Triplatin complexes.
The phosphate clamp: sequence selective nucleic acid binding profiles and conformational induction of endonuclease inhibition by cationic Triplatin complexes.
Nucleic acids research (2014-11-22)
Andreea Prisecaru, Zara Molphy, Ralph G Kipping, Erica J Peterson, Yun Qu, Andrew Kellett, Nicholas P Farrell
ABSTRACT
The substitution-inert polynuclear platinum(II) complex (PPC) series, [{trans-Pt(NH3)2(NH2(CH2)nNH3)}2-μ-(trans-Pt(NH3)2(NH2(CH2)nNH2)2}](NO3)8, where n = 5 (AH78P), 6 (AH78 TriplatinNC) and 7 (AH78H), are potent non-covalent DNA binding agents where nucleic acid recognition is achieved through use of the 'phosphate clamp' where the square-planar tetra-am(m)ine Pt(II) coordination units all form bidentate N-O-N complexes through hydrogen bonding with phosphate oxygens. The modular nature of PPC-DNA interactions results in high affinity for calf thymus DNA (Kapp ∼5 × 10(7) M(-1)). The phosphate clamp-DNA interactions result in condensation of superhelical and B-DNA, displacement of intercalated ethidium bromide and facilitate cooperative binding of Hoechst 33258 at the minor groove. The effect of linker chain length on DNA conformational changes was examined and the pentane-bridged complex, AH78P, was optimal for condensing DNA with results in the nanomolar region. Analysis of binding affinity and conformational changes for sequence-specific oligonucleotides by ITC, dialysis, ICP-MS, CD and 2D-(1)H NMR experiments indicate that two limiting modes of phosphate clamp binding can be distinguished through their conformational changes and strongly suggest that DNA condensation is driven by minor-groove spanning. Triplatin-DNA binding prevents endonuclease activity by type II restriction enzymes BamHI, EcoRI and SalI, and inhibition was confirmed through the development of an on-chip microfluidic protocol.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Water, deionized
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
SAFC
HEPES
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Supelco
Water, for TOC analysis
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Water, tested according to Ph. Eur.