  • Search Results
  • Skin regeneration effect and chemical composition of essential oil from Artemisia Montana.
Skin regeneration effect and chemical composition of essential oil from Artemisia Montana.
Natural product communications (2014-12-24)
Mi-So Yoon, Kyung-Jong Won, Do Yoon Kim, Dae Il Hwang, Seok Won Yoon, Bokyung Kim, Hwan Myung Lee
ABSTRACT
Artemisia montana Pampan (Compositae) (AMP) contains various compounds, including phenolic acids, alkaloids, and essential oil. It has been widely used in oriental medicine due to a variety of biological effects. However, the biological activity of the essential oil from AMP (AMPEO) on skin has not been investigated. In the present study, AMPEO was evaluated for its composition and its effect on cellular events (migration and proliferation) related to skin regeneration using normal human keratinocytes (HaCats). AMPEO, which was extracted by steam distillation, contained 42 components. AMPEO increased proliferation in HaCats in a dose-dependent manner (EC 50, 8.5 ng/mL) and did not affect migration. AMPEO also enhanced the phosphorylation of Akt and ERK 1/2 and induced the synthesis of type IV collagen, but not type I collagen in HaCats. In addition, AMPEO promoted wound closure in the dorsal side skin of rat tail. These results demonstrated that AMPEO extracted by steam distillation induced proliferation and synthesis of type IV collagen in human skin keratinocytes, and may thereby exert positive effects on skin regeneration and wound healing in human skin.
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-AKT1 (pThr450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MAPK3 (Ab-202) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MAPK3 (Ab-204) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody