  • Search Results
  • Assay reproducibility and interindividual variation for 15 serum estrogens and estrogen metabolites measured by liquid chromatography-tandem mass spectrometry.
Assay reproducibility and interindividual variation for 15 serum estrogens and estrogen metabolites measured by liquid chromatography-tandem mass spectrometry.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2014-12-05)
Barbara J Fuhrman, Xia Xu, Roni T Falk, Cher M Dallal, Timothy D Veenstra, Larry K Keefer, Barry I Graubard, Louise A Brinton, Regina G Ziegler, Gretchen L Gierach
ABSTRACT
Interindividual differences in estrogen metabolism may partially account for differences in risks of estrogen-responsive cancers. We conducted a proof-of-performance study to assess the reproducibility of a LC/MS-MS method for measurement of 15 serum estrogens and metabolites (all 15 termed EM) in total (conjugated+unconjugated) and unconjugated forms and describe interindividual variation. Interindividual variation in serum EM profiles was evaluated for 20 premenopausal women, 15 postmenopausal women, and 10 men. Replicate aliquots from 10 premenopausal women, 5 postmenopausal women, and 5 men were assayed eight times over 4 weeks. Components of variance were used to calculate coefficients of variation (CV) and intraclass correlation coefficients (ICC). In postmenopausal women and men, median EM concentrations were similar and substantially lower than that in premenopausal women. Within each sex/menopausal group, the sum of all EM varied 5- to 7-fold across extreme deciles. Some EM had greater variation; total estrone varied approximately 12-fold in premenopausal and postmenopausal women. Unconjugated estradiol varied 17-fold in postmenopausal women but only 5-fold in premenopausal women and men. CVs reflecting variation across replicate measures for individuals were <5% for most EM, but higher in some individuals with a low EM concentration. Overall laboratory CVs for all but one EM were <2% and ICCs were >99% for all EM in each group. The serum EM assay has excellent laboratory reproducibility. In premenopausal women, postmenopausal women, and men, interindividual variation in EM measures is substantially greater than laboratory variation. The serum EM assay is suitable for epidemiologic application. See all the articles in this CEBP Focus section, "Biomarkers, Biospecimens, and New Technologies in Molecular Epidemiology."
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dansyl chloride, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Methoxyestradiol, powder
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Dichloromethane, analytical standard, stabilized
Supelco
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Dichloromethane, ≥99.9%