- Renalase protects against contrast-induced nephropathy in Sprague-Dawley rats.
Renalase protects against contrast-induced nephropathy in Sprague-Dawley rats.
Contrast-induced nephropathy (CIN) is the third leading cause of hospital-acquired acute renal failure. Oxidative stress, apoptosis and inflammation play crucial roles in CIN. Renalase is a newly discovered monoamine oxidase from the kidney. We hypothesize that renalase could protect against CIN through anti-oxidation, anti-inflammation and anti-apoptosis pathways. We tested our hypothesis in vivo with a rat model of Ioversol-induced CIN and in vitro. Sprague-Dawley rats were divided into 4 groups (n = 6 per group): control group, Ioversol group (rats subjected to Ioversol-induced CIN), Ioversol plus vehicle group (CIN rats pretreated with vehicle) and Ioversol plus renalase group (CIN rats pretreated with 2 mg/kg recombinant renalase). HK2 cells were treated with Ioversol or H2O2. The results showed that pretreatment with renalase attenuated the deterioration of renal function, tubular necrosis, oxidative stress, apoptosis and inflammation (P<0.05). Furthermore, renalase protected HK2 cells against the cytotoxicity of Ioversol and suppressed Caspase-3 activity, oxidative stress and apoptosis induced by H2O2. Recombinant renalase protected CIN in rats through anti-oxidation, anti-apoptosis and anti-inflammation mechanisms.