  • Search Results
  • Carbonic anhydrase XII is a new therapeutic target to overcome chemoresistance in cancer cells.
Carbonic anhydrase XII is a new therapeutic target to overcome chemoresistance in cancer cells.
Oncotarget (2015-02-18)
Joanna Kopecka, Ivana Campia, Andrea Jacobs, Andreas P Frei, Dario Ghigo, Bernd Wollscheid, Chiara Riganti
ABSTRACT
Multidrug resistance (MDR) in cancer cells is a challenging phenomenon often associated with P-glycoprotein (Pgp) surface expression. Finding new ways to bypass Pgp-mediated MDR still remains a daunting challenge towards the successful treatment of malignant neoplasms such as colorectal cancer.We applied the Cell Surface Capture technology to chemosensitive and chemoresistant human colon cancer to explore the cell surface proteome of Pgp-expressing cells in a discovery-driven fashion. Comparative quantitative analysis of identified cell surface glycoproteins revealed carbonic anhydrase type XII (CAXII) to be up-regulated on the surface of chemoresistant cells, similarly to Pgp. In cellular models showing an acquired MDR phenotype due to the selective pressure of chemotherapy, the progressive increase of the transcription factor hypoxia-inducible factor-1 alpha was paralleled by the simultaneous up-regulation of Pgp and CAXII. CAXII and Pgp physically interacted at the cell surface. CAXII silencing or pharmacological inhibition with acetazolamide decreased the ATPase activity of Pgp by altering the optimal pH at which Pgp operated and promoted chemosensitization to Pgp substrates in MDR cells.We propose CAXII as a new secondary marker of the MDR phenotype that influences Pgp activity directly and can be used as a pharmacological target for MDR research and potential treatment.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
SAFC
Iodoacetamide
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
2,2′-Thiodiethanol, ≥99%
Sigma-Aldrich
2,2′-Thiodiethanol, purum, ≥95.0% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)