  • Search Results
  • Reduction of plasma aldosterone and arterial stiffness in obese pre- and stage1 hypertensive subjects after aerobic exercise.
Reduction of plasma aldosterone and arterial stiffness in obese pre- and stage1 hypertensive subjects after aerobic exercise.
Journal of human hypertension (2014-05-03)
S R Collier, K Sandberg, A M Moody, V Frechette, C D Curry, H Ji, R Gowdar, D Chaudhuri, M Meucci
ABSTRACT
Obesity-related hypertension is associated with increased activity of the renin-angiotensin-aldosterone system (RAAS), increasing arterial stiffness. Aerobic exercise decreases pulse wave velocity (PWV), therefore a treatment option for hypertension and obesity. Assess RAAS activity and PWV before and after 4 weeks of aerobic training in unmedicated, pre-to-stage-1 hypertensives. Ten obese subjects (52±3.2 years, body mass index=33.5±1.4) performed 30 min of aerobic exercise on a treadmill 3 days per week at 65% of peak oxygen consumption (VO2peak). Descriptive characteristics, systolic and diastolic blood pressure (SBP and DBP), PWV, and a blood draw was performed at baseline, following the 4-week control and training interventions. No differences in descriptive characteristics during the control period were observed, however, a significant decrease in plasma aldosterone (ALDO) (255.4±75 to 215.8±66 pg ml(-1), P=0.001), SBP (140±12 to 136±10.4 mm Hg; P=0.02), DBP (89±4.2 to 85±6.3 mm Hg; P=0.03) and central PWV (11.2±0.6 to 9.8±0.8 m s(-1); P=0.04) was shown pre-to-post exercise training. Four weeks of moderate-intensity aerobic training in obese, hypertensives decreases plasma ALDO independently of body weight and is significantly correlated to decreases in PWV reductions.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)