  • Search Results
  • Investigation of cytotoxicity of phosphoryl choline modified single-walled carbon nanotubes under a live cell station.
Investigation of cytotoxicity of phosphoryl choline modified single-walled carbon nanotubes under a live cell station.
BioMed research international (2014-07-25)
Yufeng Zhao, Qunlong Mao, Yu Liu, Yan Zhang, Tao Zhang, Zhengsheng Jiang
ABSTRACT
Single-walled carbon nanotubes (SWCNTs) and various modified SWCNTs have drawn a lot of attention due to their potential applications in biomedical field. Before further moving on to real clinical applications, hydrophobicity and toxicity of SWCNTs should be investigated thoroughly. In this paper, 2-methacryloyloxy ethyl phosphorylcholine (MPC) was adopted to modify SWCNTs and phosphoryl choline was grafted onto SWCNTs as small molecule moieties and polymeric chains, which made SWCNTs dispersed stably both in water and in cell culture medium for a long time. Cytotoxicity of pristine and modified SWCNTs were assayed upon successful preparation of the designed modified SWCNT. Furthermore, the internalization of SWCNTs by three cells was investigated using a live cell station under normal culture temperature (37°C) and low temperature (4°C). The results showed that the internalization of modified SWCNTs was related to both the active transport and the passive transport. Although the modification with phosphoryl choline remarkably reduced the cytotoxicity of SWCNTs, the results were probably due to other reasons such as the decrease in the ratio of cells which internalized modified SWCNTs since the cells without SWCNTs occupation still exhibited normal states.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Thionyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture