  • Search Results
  • Induction of dental pulp-derived induced pluripotent stem cells in the absence of c-Myc for differentiation into neuron-like cells.
Induction of dental pulp-derived induced pluripotent stem cells in the absence of c-Myc for differentiation into neuron-like cells.
Journal of the Chinese Medical Association : JCMA (2014-12-03)
Yun-Ching Chang, Wan-Chun Li, Nae-Fang Twu, Hsin-Yang Li, Wen-Liang Lo, Yuh-Lih Chang, Yi-Yen Lee, Chun-Fu Lin, Yang-Hsin Shih, Ming-Teh Chen
ABSTRACT
A recent research breakthrough has demonstrated that the ectopic expression of four genes is sufficient to reprogram human fibroblasts into inducible pluripotent stem cells (iPSCs). However, whether human dental pulp cells (DPCs) could be reprogrammed into iPSCs remains an open question. In this study, we demonstrated that DPCs from deciduous and permanent teeth can be reprogrammed into iPSCs without c-Myc and had the capacity to differentiate into neuron-like cells. DPCs were obtained from donors and reprogrammed into iPSCs using retroviral transduction with SOX2, OCT4, and KLF4. Then, these iPSCs were differentiated into neuron-like cells. Microarray and bioinformatics were used to compare the gene expression profile among these iPSCs and iPSC-derived neuron-like cells. The DPCs displayed a high vitality and capability to quickly restart proliferation and expressed elevated pluripotency similar to mesenchymal stem cells. According to our results, DPC-derived iPSC colonies that could be subcultured and propagated were established as early as 10 days after transduction, in comparison with the skin fibroblast (DPC-derived iPSCs) without c-Myc presented embryonic stem cell-like properties and the pluripotent potential to differentiate into neuron-like cells, which resemble neurons both morphologically and functionally. The human DPCs from deciduous and permanent teeth can undergo reprogramming to establish pluripotent stem cell lines without c-Myc. These surgical residues, usually regarded as medical waste, can be used as an alternative source of pluripotent stem cells for personalized medicine.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
HEPES
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
1,4-Diaminobutane, 99%
SAFC
L-Glutamine
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Putrescine, analytical standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
SAFC
HEPES
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture