  • Search Results
  • Antioxidant and antiproliferative activities of twenty-four Vitis vinifera grapes.
Antioxidant and antiproliferative activities of twenty-four Vitis vinifera grapes.
PloS one (2014-08-19)
Zhenchang Liang, Lailiang Cheng, Gan-Yuan Zhong, Rui Hai Liu
ABSTRACT
Grapes are rich in phytochemicals with many proven health benefits. Phenolic profiles, antioxidant and antiproliferative activities of twenty-four selected Vitis vinifera grape cultivars were investigated in this study. Large ranges of variation were found in these cultivars for the contents of total phenolics (95.3 to 686.5 mg/100 g) and flavonoids (94.7 to 1055 mg/100 g) and antioxidant activities (oxygen radical absorbance capacity 378.7 to 3386.0 mg of Trolox equivalents/100 g and peroxylradical scavenging capacity14.2 to 557 mg of vitamin C equivalents/100 g), cellular antioxidant activities (3.9 to 139.9 µmol of quercetin equivalents/100 g without PBS wash and 1.4 to 95.8 µmol of quercetin equivalents /100 g with PBS wash) and antiproliferative activities (25 to 82% at the concentrations of 100 mg/mL extracts).The total antioxidant activities were significantly correlated with the total phenolics and flavonoids. However, no significant correlations were found between antiproliferative activities and total phenolics or total flavonoids content. Wine grapes and color grapes showed much higher levels of phytochemicals and antioxidant activities than table grapes and green/yellow grapes. Several germplasm accessions with much high contents of phenolics and flavonoids, and total antioxidant activity were identified. These germplasm can be valuable sources of genes for breeding grape cultivars with better nutritional qualities of wine and table grapes in the future.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%