  • Search Results
  • Alkaline cation-exchange chromatography for the reduction of aggregate and a mis-formed disulfide variant in a bispecific antibody purification process.
Alkaline cation-exchange chromatography for the reduction of aggregate and a mis-formed disulfide variant in a bispecific antibody purification process.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
Troii Hall, Joseph J Wilson, Tammy J Brownlee, James R Swartling, Sarah E Langan, Peter K Lambooy
ABSTRACT
During the purification development of a bispecific antibody, cation-exchange chromatography was screened for its ability to separate a prominently expressed (>12%) mis-formed disulfide bond variant, termed MAb-diabody, and aggregate from the product of interest. The influence of pH, product load (g of product per liter of resin) and linear velocity on the separations were evaluated for the strong cation-exchange resins SP Sepharose HP and POROS(®) HS50. Cation-exchange chromatography is commonly operated distant to the isoelectric point of a molecule, generally leading to acidic conditions for antibody purification. However, the results herein demonstrated improved removal of MAb-diabody with increasing pH, resulting in reduction of MAb-diabody content greater than 12-fold when operating near the alkaline pI of the product. This approach was successful over a range of linear velocities and g/L of resin loading. Aggregate removal was less affected by pH and was effectively reduced from 10.9% to less than 3% for each condition. Furthermore, this method was successfully scaled to a 60 cm diameter column using SP Sepharose HP resin.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
BICINE, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
CAPSO, ≥99% anhydrous basis (titration)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride solution, 0.85%