  • Search Results
  • 19F NMR as a probe of ligand interactions with the iNOS binding site of SPRY domain-containing SOCS box protein 2.
19F NMR as a probe of ligand interactions with the iNOS binding site of SPRY domain-containing SOCS box protein 2.
Chemical biology & drug design (2014-05-13)
Eleanor W W Leung, Hiromasa Yagi, Jitendra R Harjani, Mark D Mulcair, Martin J Scanlon, Jonathan B Baell, Raymond S Norton
ABSTRACT
SPRY domain-containing SOCS box protein 2 (SPSB2) regulates inducible nitric oxide synthase (iNOS) by targeting it for proteasomal degradation. Inhibiting this interaction prolongs the intracellular lifetime of iNOS, leading in turn to enhanced killing of infectious pathogens such as bacteria and parasites. SPSB2 recognizes a linear motif (DINNN) in the disordered N-terminus of iNOS, and ligands that target the DINNN binding site on SPSB2 are potentially novel anti-infective agents. We have explored (19)F NMR as a means of probing ligand binding to SPSB2. All six Trp residues in SPSB2 were replaced with 5-fluorotryptophan (5-F-Trp) by utilizing a Trp auxotroph strain of Escherichia coli. The labeled protein was well folded and bound a DINNN-containing peptide with similar affinity to native SPSB2. Six well-resolved 5-F-Trp resonances were observed in the (19)F NMR spectrum and were assigned using site-directed mutagenesis. The (19)F resonance of W207 was significantly perturbed upon binding to DINNN-containing peptides. Other resonances were perturbed to a lesser extent although in a way that was sensitive to the composition of the peptide. Analogues of compounds identified in a fragment screen also perturbed the W207 resonance, confirming their binding to the iNOS peptide-binding site on SPSB2. (19)F NMR promises to be a valuable approach in developing inhibitors that bind to the DINNN binding site.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)