  • Search Results
  • Highly sensitive and selective H2 sensing by ZnO nanofibers and the underlying sensing mechanism.
Highly sensitive and selective H2 sensing by ZnO nanofibers and the underlying sensing mechanism.
Journal of hazardous materials (2015-01-16)
Akash Katoch, Sun-Woo Choi, Hyoun Woo Kim, Sang Sub Kim
ABSTRACT
We report, and propose a mechanism for, the exceptional hydrogen gas (H2) sensing ability of ZnO nanofibers. In comparison to SnO2 nanofibers, ZnO nanofibers show outstanding H2 gas response and unmistakable H2 selectivity. Different from the reducing gas effect observed in SnO2 nanofibers, a semiconductor-to-metal transition that occurs in the presence of H2 gas molecules is responsible for the exceptional response and selectivity of ZnO nanofibers to H2. Notably, the presence of nanograins within nanofibers further intensifies the resistance modulation observed due to this transition.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Tin(II) chloride dihydrate, ACS reagent, 98%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Tin(II) chloride dihydrate, reagent grade, 98%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.995% trace metals basis
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Tin(II) chloride dihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Tin(II) chloride dihydrate, suitable for AAS
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.99% trace metals basis
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard