  • Search Results
  • Cutting edge: CD69 interference with sphingosine-1-phosphate receptor function regulates peripheral T cell retention.
Cutting edge: CD69 interference with sphingosine-1-phosphate receptor function regulates peripheral T cell retention.
Journal of immunology (Baltimore, Md. : 1950) (2015-01-28)
Laura K Mackay, Asolina Braun, Bethany L Macleod, Nicholas Collins, Christina Tebartz, Sammy Bedoui, Francis R Carbone, Thomas Gebhardt
ABSTRACT
Tissue-resident memory T cells provide local immune protection in barrier tissues, such as skin and mucosa. However, the molecular mechanisms controlling effector T cell retention and subsequent memory formation in those locations are not fully understood. In this study, we analyzed the role of CD69, an early leukocyte activation marker, in regulating effector T cell egress from peripheral tissues. We provide evidence that CD69 surface expression by skin-infiltrating CD8 T cells can be regulated at multiple levels, including local Ag stimulation and signaling through type I IFNRs, and it coincides with the transcriptional downregulation of the sphingosine-1-phosphate receptor S1P1. Importantly, we demonstrate that expression of CD69, by interfering with sphingosine-1-phosphate receptor function, is a critical determinant of prolonged T cell retention and local memory formation. Our results define an important step in the generation of long-lived adaptive immune memory at body surfaces.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, purum p.a., ≥98.0% (GC)
Supelco
1-Fluoro-2,4-dinitrobenzene, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium (meta)periodate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Sodium periodate, 10 wt. %, matrix silica gel support