MilliporeSigma
Urinary prostaglandin E2 metabolite and breast cancer risk.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2014-09-13)
Yong Cui, Xiao-Ou Shu, Yu-Tang Gao, Qiuyin Cai, Bu-Tian Ji, Hong-Lan Li, Nathaniel Rothman, Jie Wu, Gong Yang, Yong-Bing Xiang, Wei Zheng
ABSTRACT
Levels of the cyclooxygenase 2 (COX2) enzyme are elevated in breast cancer tissue, and most COX2 effects are believed to be mediated through overproduction of prostaglandin E2 (PGE2). We evaluated associations between the primary urinary metabolite of PGE2 (PGE-M) and breast cancer risk. A nested case-control study of 504 cases and 1,082 controls was conducted using data from the Shanghai Women's Health Study, a large population-based prospective cohort study of 74,941 Chinese women. Urinary PGE-M was measured using a liquid chromatography/tandem mass spectrometric method. Logistic regression estimated odds ratios (OR) and 95% confidence intervals (95% CI) with adjustment for potential confounders. Overall, no association between urinary PGE-M and breast cancer was detected. However, a suggestive positive association was found among postmenopausal women. In particular, a clear dose-response relationship between urinary PGE-M and breast cancer was observed among postmenopausal women with a body mass index (BMI) < 25 kg/m(2) (Plinear trend = 0.005). Among these women, risk of breast cancer increased from 1.00 (reference) to 1.06 (95% CI, 0.56-1.99), 1.50 (95% CI, 0.79-2.83), and 2.32 (95% CI, 1.24-4.41) for the lowest to highest quartiles of PGE-M, and such associations were stronger among those who were diagnosed with cancer within the first four years of sample collection. No apparent association was observed among overweight postmenopausal women (BMI ≥ 25 kg/m(2)). High urinary PGE-M level was associated with elevated risk of breast cancer among normal weight, postmenopausal women. Urinary PGE-M level may be useful for breast cancer risk assessment among normal weight, postmenopausal women. Cancer Epidemiol Biomarkers Prev; 23(12); 2866-73. ©2014 AACR.
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%