  • Search Results
  • Fruits and barks extracts of Zanthozyllum heitzii a spice from Cameroon induce mitochondrial dependent apoptosis and Go/G1 phase arrest in human leukemia HL-60 cells.
Fruits and barks extracts of Zanthozyllum heitzii a spice from Cameroon induce mitochondrial dependent apoptosis and Go/G1 phase arrest in human leukemia HL-60 cells.
Biological research (2015-02-28)
Constant Anatole Pieme, Guru Kumar Santosh, Emmanuel Mouafo Tekwu, Tülin Askun, Hatice Aydeniz, Jeanne Yonkeu Ngogang, Shashi Bhushan, Ajit Kumar Saxena
ABSTRACT
Zanthoxylum heitzii is a spice used to prepare several dishes and to treat tumors, syphilis, malaria, cardiac palpitations, urogenital infections in the west region of Cameroon, but the antitumor mechanisms and chemical composition are not yet investigated. This study was aimed to determine the antiproliferative effects of four extracts from the fruits and barks of Zanthoxyllum heitzii (Rutaceae) on apoptosis in human promyelocytic cells, their mechanisms and the chemical composition. The 3-(4, 5-dimethylthiazole-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay was used to determine the fifty percent inhibition (IC50) concentration of the cell lines after treatment. The effect on morphology was observed using a light or fluorescence microscopy. The rate of apoptosis and the cell cycle were measured using flow cytometry (FCM). The phytochemical analysis of the extract was carried with HPLC/MS methods. The phytochemical analysis of the extracts indicated the presence of four known polyphenols (Syringic acid, Juglon, Luteolin and Myricetin) in both fruits and barks of Z. heitzii but in different quantities. Syringic acid and Myricetin concentrations were between 17-21 fold higher in the fruits than the stem bark. Rhamnetin (393.35 μg/mL) and Oleuropein (63.10 μg/mL) were identified only in the stem barks of Z. heitzii. Among the four extracts tested for cytotoxicity properties, only the methanol extract of fruits and barks significantly inhibited cell proliferation of HL-60 cells with IC50 value of 20 μg/mL and 12 μg/mL respectively. HL-60 cells treated with Z. heitzii extracts significantly produced reactive oxygen species (ROS) with concurrent loss of mitochondrial membrane potential (MMP). Modifications in the DNA distribution and enhanced of G1/G0 phase cell cycle arrest were observed in a concentration dependent manner. Polyphenols from Z. heitzii plant exert inhibitory effect on HL-60 cells through the reactive oxygen species (ROS) generation, loss of mitochondrial membrane potential and cell cycle destabilization.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Gallic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Rosmarinic acid, 96%