  • Search Results
  • A validated method for quantifying hypoglycin A in whole blood by UHPLC-HRMS/MS.
A validated method for quantifying hypoglycin A in whole blood by UHPLC-HRMS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-23)
Jérémy Carlier, Jérôme Guitton, Cécile Moreau, Baptiste Boyer, Fabien Bévalot, Laurent Fanton, Jean Habyarimana, Gilbert Gault, Yvan Gaillard
ABSTRACT
Hypoglycin A (HGA) is the toxic principle in ackee (Blighia sapida Koenig), a nutritious and readily available fruit which is a staple of the Jamaican working-class and rural population. The aril of the unripe fruit has high concentrations of HGA, the cause of Jamaican vomiting sickness, which is very often fatal. HGA is also present in the samara of several species of maple (Acer spp.) which are suspected to cause seasonal pasture myopathy in North America and equine atypical myopathy in Europe, often fatal for horses. The aim of this study was to develop a method for quantifying HGA in blood that would be sensitive enough to provide toxicological evidence of ackee or maple poisoning. Analysis was carried out using solid-phase extraction (HILIC cartridges), dansyl derivatization and UHPLC-HRMS/MS detection. The method was validated in whole blood with a detection limit of 0.35 μg/L (range: 0.8-500 μg/L). This is the first method applicable in forensic toxicology for quantifying HGA in whole blood. HGA was quantified in two serum samples from horses suffering from atypical myopathy. The concentrations were 446.9 and 87.8 μg/L. HGA was also quantified in dried arils of unripe ackee fruit (Suriname) and seeds of sycamore maple (Acer pseudoplatanus L.) (France). The concentrations were 7.2 and 0.74 mg/g respectively.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)