  • Search Results
  • Proteomic studies of pediatric medulloblastoma tumors with 17p deletion.
Proteomic studies of pediatric medulloblastoma tumors with 17p deletion.
Journal of proteome research (2014-12-30)
Athanasios K Anagnostopoulos, Chrissa Papathanassiou, Kalliopi Karamolegou, Ema Anastasiadou, Konstantinos S Dimas, Harry Kontos, Anastasios Koutsopoulos, Neofytos Prodromou, Fotini Tzortzatou-Stathopoulou, George Th Tsangaris
ABSTRACT
CNS tumors are the leading cause of cancer-related death in children. Medulloblastoma is the commonest pediatric CNS malignancy, wherein, despite multimodal therapy with surgery, radiation, and chemotherapy, 5 year survival rates merely approach 60%. Until present, gene expression and cytogenetic studies have produced contradicting findings regarding the molecular background of the specific disease. Through integration of genomics, bioinformatics, and proteomics, the current study aims to shed light at the proteomic-related molecular events responsible for MBL pathophysiology, as well as to provide molecular/protein/pathway answers concerning tumor-onset. Experiments were performed on tissues collected at surgery. With 17p loss being the commonest chromosomal aberrance observed in our sample set, array-CGH were employed to first distinguish for 17p-positive cases. 2-DE coupled to mass spectrometry identification exposed the MBL-specific protein profile. Protein profiles of malignant tissues were compared against profiles of normal cerebellar tissues, and quantitative protein differences were determined. Bioinformatics, functional and database analyses, characterization, and subnetwork profiling generated information on MBL protein interactions. Key molecules of the PI3K/mTOR signaling network were identified via the techniques applied herein. Among the findings IGF2, PI3K, Rictor, MAPKAP1, S6K1, 4EBP1, and ELF4A, as part of the IGF network (implicating PI3K/mTOR), were founded to be deregulated.
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Sucrose, analytical standard
Urea, European Pharmacopoeia (EP) Reference Standard