  • Search Results
  • Electrochemistry of moexipril: experimental and computational approach and voltammetric determination.
Electrochemistry of moexipril: experimental and computational approach and voltammetric determination.
Die Pharmazie (2014-10-03)
Hüdai I Taşdemir, E Kiliç
ABSTRACT
The electrochemistry of moexipril (MOE) was studied by electrochemical methods with theoretical calculations performed at B3LYP/6-31 + G (d)//AM1. Cyclic voltammetric studies were carried out based on a reversible and adsorption-controlled reduction peak at -1.35 V on a hanging mercury drop electrode (HMDE). Concurrently irreversible diffusion-controlled oxidation peak at 1.15 V on glassy carbon electrode (GCE) was also employed. Potential values are according to Ag/AgCI, (3.0 M KCI) and measurements were performed in Britton-Robinson buffer of pH 5.5. Tentative electrode mechanisms were proposed according to experimental results and ab-initio calculations. Square-wave adsorptive stripping voltammetric methods have been developed and validated for quantification of MOE in pharmaceutical preparations. Linear working range was established as 0.03-1.35 microM for HMDE and 0.2-20.0 microM for GCE. Limit of quantification (LOQ) was calculated to be 0.032 and 0.47 microM for HMDE and GCE, respectively. Methods were successfully applied to assay the drug in tablets by calibration and standard addition methods with good recoveries between 97.1% and 106.2% having relative standard deviation less than 10%.
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
SAFC
Acetic acid, glacial
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C