  • Search Results
  • Pretreatment with buthionine sulfoximine enhanced uptake and retention of BSH in brain tumor.
Pretreatment with buthionine sulfoximine enhanced uptake and retention of BSH in brain tumor.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2014-04-16)
F Yoshida, T Yamamoto, K Nakai, A Zaboronok, M Matsuda, H Akutsu, E Ishikawa, M Shirakawa, A Matsumura
ABSTRACT
To determine the influence of buthionine sulfoximine (BSO) on boron biodistribution after sulfhydryl borane (BSH) administration for boron neutron capture therapy, the effectiveness of the combination of BSO with sulfhydril- (BSH) and non-sulfhydril (B12H12 and BNH3) boron compounds, and the interval between BSO and BSH administration, the retention of boron in tissues have been evaluated using a 9L rat tumor model. Simultaneous administration of BSH and BSO showed significantly higher boron accumulation compared to that without BSO, however there was no difference in tissue boron level between B12H12 and BNH3 administration with BSO or without BSO. The longer interval (6h) between BSH and BSO administration related to the highest boron concentration in the brain and subcutaneous tumors compared to shorter intervals (0.5, 3h). Boron concentration in subcutaneous and brain tumors was maintained for 6 and 12h after the administration of BSH following BSO pretreatment.
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Sigma-Aldrich
Boron, crystalline, −60 mesh, 99% trace metals basis
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Boron, monofilament, 5m, diameter 0.1mm
Boron, microfoil, disks, 25mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
Boron, monofilament, 20m, diameter 0.2mm
Boron, monofilament, 200m, diameter 0.1mm
Boron, monofilament, 50m, diameter 0.2mm
Boron, monofilament, 20m, diameter 0.1mm
Boron, monofilament, 5m, diameter 0.2mm
Boron, monofilament, 100m, diameter 0.1mm
Boron, monofilament, 10m, diameter 0.1mm
Boron, monofilament, 50m, diameter 0.1mm
Boron, monofilament, 10m, diameter 0.2mm
Boron, microfoil, disks, 10mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%