  • Search Results
  • The age factor for mitoxantrone's cardiotoxicity: multiple doses render the adult mouse heart more susceptible to injury.
The age factor for mitoxantrone's cardiotoxicity: multiple doses render the adult mouse heart more susceptible to injury.
Toxicology (2015-01-15)
José Luís Dores-Sousa, José Alberto Duarte, Vítor Seabra, Maria de Lourdes Bastos, Félix Carvalho, Vera Marisa Costa
ABSTRACT
Age is a known susceptibility factor for the cardiotoxicity of several anticancer drugs, including mitoxantrone (MTX). The impact of anticancer drugs in young patients is underestimated, thus we aimed to evaluate the cardiotoxicity of MTX in juvenile and adult animals. Juvenile (3 week-old) and adult (8-10 week-old) male CD-1 mice were used. Each group was treated with a 9.0mg/kg cumulative dose of MTX or saline; they were maintained in a drug-free period for 3-weeks after the last administration to allow the development of late toxicity (protocol 1), or sacrificed 24h after the last MTX administration to evaluate early cardiotoxicity (protocol 2). In protocol 1, no adult mice survived, while 2 of the juveniles reached the end of the protocol. High plasma aspartate aminotransferase/alanine aminotransferase ratio and a high cardiac reduced/oxidized glutathione ratio were found in the surviving MTX-treated juvenile mice. In protocol 2, a significant decrease in plasma creatine-kinase MB in juveniles was found 24h after the last MTX-administration. Cardiac histology showed that both MTX-treated populations had significant damage, although higher in adults. However, MTX-treated juveniles had a significant increase in fibrotic tissue. The MTX-treated adults had higher values of cardiac GSSG and protein carbonylation, but lower cardiac noradrenaline levels. For the first time, mature adult animals were shown to be more susceptible to MTX as evidenced by several biomarkers, while young animals appear to better adjust to the MTX-induced cardiac injury. Even so, the higher level of fibrotic tissue and the histological damage showed that MTX also causes cardiac damage in the juvenile population.
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)