  • Search Results
  • Protective effect of Aronia melanocarpa polyphenols against cadmium-induced disorders in bone metabolism: a study in a rat model of lifetime human exposure to this heavy metal.
Protective effect of Aronia melanocarpa polyphenols against cadmium-induced disorders in bone metabolism: a study in a rat model of lifetime human exposure to this heavy metal.
Chemico-biological interactions (2015-02-07)
Malgorzata M Brzóska, Joanna Rogalska, Malgorzata Galazyn-Sidorczuk, Maria Jurczuk, Alicja Roszczenko, Michal Tomczyk
ABSTRACT
It was investigated, in a female rat model of low and moderate lifetime human exposure to cadmium (Cd), whether polyphenols from Aronia melanocarpa berries (chokeberry; AMP) may offer protection from this heavy metal-induced disorders in bone metabolism. For this purpose, numerous indices of bone formation (osteocalcin, alkaline phosphatase, osteoprotegerin) and resorption (carboxy-terminal cross-linking telopeptides of type I collagen, soluble receptor activator of nuclear factor-κB ligand) in the serum and/or distal femur epiphysis (trabecular bone region), as well as bone mineral status (volumetric bone mineral density of the femur and content of mineral components, including calcium, in the bone tissue at the distal femur epiphysis) were evaluated in female Wistar rats that received a 0.1% aqueous extract of AMP, as the only drinking fluid (prepared from lyophilized extract by Adamed Consumer Healthcare), and/or Cd in diet (1 and 5mg/kg) for 3, 10, 17, and 24 months. Examination of the phytochemical profile of the aronia extract revealed high content of polyphenols (612.40 ± 3.33 mg/g), including anthocyanins, proanthocyanidins, phenolic acids, and flavonoids. Among detected compounds anthocyanins were identified as dominating. The exposure to Cd, dose- and duration-dependently, enhanced resorption and inhibited formation of the bone tissue resulting in its decreased mineralization. The administration of AMP under the exposure to 1 and 5 mgCd/kg diet provided important protection from this heavy metal-induced disturbances in the bone turnover and changes in the bone mineral status, and the beneficial impact of polyphenols resulted from their independent action and interaction with Cd. These findings suggest that consumption of Aronia melanocarpa polyphenols may play a role in prevention against female skeleton damage due to chronic exposure to Cd and that chokeberry represents the good natural plant candidate for further investigations of its prophylactic use under environmental exposure to this heavy metal.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Hexamethylenetetramine, ACS reagent, ≥99.0%
Sigma-Aldrich
Hexamethylenetetramine, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Lanthanum(III) chloride hydrate, 99.9% trace metals basis
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylenetetramine, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Supelco
Gallic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Methenamine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Hexamethylenetetramine
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)