  • Search Results
  • Molecular stereotactic biopsy technique improves diagnostic accuracy and enables personalized treatment strategies in glioma patients.
Molecular stereotactic biopsy technique improves diagnostic accuracy and enables personalized treatment strategies in glioma patients.
Acta neurochirurgica (2014-05-06)
Sabina Eigenbrod, Raimund Trabold, David Brucker, Christian Erös, Rupert Egensperger, Christian La Fougere, Werner Göbel, Adrian Rühm, Hans A Kretzschmar, Jörg C Tonn, Jochen Herms, Armin Giese, Friedrich W Kreth
ABSTRACT
In gliomas molecular biomarkers are increasingly gaining diagnostic, prognostic and predictive significance. Determination of biomarker status after biopsy is important as not all patients are eligible for open tumor resection. We developed and validated prospectively (6/10-12/11) a protocol allowing for both reliable determination of multiple biomarkers and representative histological diagnoses from small-sized biopsies. All molecular stereotactic biopsies were performed according to a detailed workflow. The selection of specimens best suited for molecular analyses was intra-operatively guided by the attending neuropathologist. Postoperative screening was done by methylation specific PCR using two distinct cryopreserved specimens to test for reproducibility of the findings and to rule out contamination. The DNA of a single best-suited specimen (1 mm(3)) was subjected to detailed molecular analysis (MGMT promoter methylation, IDH1/2 mutational status, LOH 1p and/or 19q). 159 consecutively enrolled untreated gliomas were analyzed (94 glioblastomas, 2 gliosarcomas, 24 anaplastic astrocytomas, 10 oligo-tumors grade II/III, 20 grade II astrocytomas and 9 pilocytic astrocytomas). Transient morbidity was 2 %. Overall, the drop-out rate due to tissue contamination was 0.4 %. Median time from biopsy to histological and molecular genetic analyses was 3 and 5 days, respectively. Distributions of the respective biomarker status for tumor subgroups were consistent with the literature. The final histological diagnosis was changed/modified in 5/159 patients according to molecular findings. Treatment after molecular biopsy was highly personalized. Molecular stereotactic biopsy is feasible and safe, can be implemented in daily clinical practice, improves diagnostic precision and enables personalized treatment.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, ascites fluid
Sigma-Aldrich
Anti-MAP2 antibody, Mouse Monoclonal, clone HM-2, purified from hybridoma cell culture
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Monoclonal Anti-MAP2 (2a+2b) antibody produced in mouse, clone AP-20, ascites fluid
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-MAP2 (2a+2b) antibody produced in mouse, ~2 mg/mL, clone AP-20, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C