  • Search Results
  • Certification of reference materials for the determination of alkylphenols.
Certification of reference materials for the determination of alkylphenols.
Analytical and bioanalytical chemistry (2015-02-07)
Nobuyasu Hanari, Keiichiro Ishikawa, Yoshitaka Shimizu, Satoko Otsuka, Ryoko Iwasawa, Naomi Fujiki, Masahiko Numata, Takashi Yarita, Kenji Kato
ABSTRACT
Certified reference materials (CRMs) are playing an increasingly important role in national and international standardizing activities. In Japan, primary standard solutions for analyses of endocrine disrupters are supplied under the national standards dissemination system named the Japan Calibration Service System (JCSS). For the traceability on reference materials used for preparation of the primary standard solutions based on the JCSS, the National Metrology Institute of Japan, National Institute of Advanced Industrial Science and Technology (NMIJ/AIST) has developed and certified high-purity reference materials of alkylphenols as NMIJ CRMs, such as 4-n-nonylphenol, 4-tert-octylphenol, 4-n-heptylphenol, 4-tert-butylphenol, and 2,4-dichlorophenol. Thereafter, it is essential to determine the alkylphenols by using these solutions based on the JCSS for environmental monitoring and risk assessments because analytical values obtained by using the solutions can ensure the reliability and traceability of the chemical analyses.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%