  • Search Results
  • Detailed characterization of a long-term rodent model of critical illness and recovery.
Detailed characterization of a long-term rodent model of critical illness and recovery.
Critical care medicine (2015-02-24)
Neil E Hill, Saima Saeed, Rahul Phadke, Matthew J Ellis, Darren Chambers, Duncan R Wilson, Josiane Castells, Jerome Morel, Damien G Freysennet, Stephen J Brett, Kevin G Murphy, Mervyn Singer
ABSTRACT
To characterize a long-term model of recovery from critical illness, with particular emphasis on cardiorespiratory, metabolic, and muscle function. Randomized controlled animal study. University research laboratory. Male Wistar rats. Intraperitoneal injection of the fungal cell wall constituent, zymosan or n-saline. Following intervention, rats were followed for up to 2 weeks. Animals with zymosan peritonitis reached a clinical and biochemical nadir on day 2. Initial reductions were seen in body weight, total body protein and fat, and muscle mass. Leg muscle fiber diameter remained subnormal at 14 days with evidence of persisting myonecrosis, even though gene expression of regulators of muscle mass (e.g., MAFbx, MURF1, and myostatin) had peaked on days 2-4 but normalized by day 7. Treadmill exercise capacity, forelimb grip strength, and in vivo maximum tetanic force were also reduced. Food intake was minimal until day 4 but increased thereafter. This did not relate to appetite hormone levels with early (6 hr) rises in plasma insulin and leptin followed by persisting subnormal levels; ghrelin levels did not change. Serum interleukin-6 level peaked at 6 hours but had normalized by day 2, whereas interleukin-10 remained persistently elevated and high-density lipoprotein cholesterol persistently depressed. There was an early myocardial depression and rise in core temperature, yet reduced oxygen consumption and respiratory exchange ratio with a loss of diurnal rhythmicity that showed a gradual but incomplete recovery by day 7. This detailed physiological, metabolic, hormonal, functional, and histological muscle characterization of a model of critical illness and recovery reproduces many of the findings reported in human critical illness. It can be used to assess putative therapies that may attenuate loss, or enhance recovery, of muscle mass and function.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Supelco
Hematoxylin
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Pefabloc® SC, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material