  • Search Results
  • Enhanced protection of pathogenic Escherichia coli ingested by a soil nematode Caenorhabditis elegans against sanitizer treatments.
Enhanced protection of pathogenic Escherichia coli ingested by a soil nematode Caenorhabditis elegans against sanitizer treatments.
Bioscience, biotechnology, and biochemistry (2014-07-24)
Mi Ri Park, Sangnam Oh, Hyun Sun Yun, Soon Han Kim, Young Ho Ko, Jee-Hoon Ryu, Min Suk Rhee, Ok Sarah Shin, Younghoon Kim
ABSTRACT
We employed Caenorhabditis elegans as a model to study the effectiveness of sanitizers in killing pathogenic Escherichia coli strains ingested by free-living nematodes. Adult worms that had fed on six pathogenic E. coli strains (highly persistent in the nematode intestine) were treated with three chemical solutions. In planktonic cells, none of the H2O2 and acetic acid treatments influenced the survival of the pathogenic E. coli strains, whereas sodium hypochlorite critically decreased the viability of the strains. Importantly, the survival of the E. coli strains was dramatically increased by persistence in the C. elegans gut under 0.1% sodium hypochlorite, and several strains could survive at a concentration of 0.5%. In addition, all pathogenic E. coli strains in the C. elegans gut survived on the lettuce for 5 days even though they were washed with 0.1% sodium hypochlorite. Taken together, our results indicate that pathogenic E. coli ingested by C. elegans may be protected against washing treatment with commercial sanitizers on raw food materials.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 10-15 %
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 4.00-4.99 %
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Sodium hypochlorite solution, 6-14% active chlorine basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis