  • Search Results
  • Assessment of low-dose cisplatin as a model of nausea and emesis in beagle dogs, potential for repeated administration.
Assessment of low-dose cisplatin as a model of nausea and emesis in beagle dogs, potential for repeated administration.
Experimental brain research (2014-05-06)
Hannah Kenward, Ludovic Pelligand, Jonathan Elliott
ABSTRACT
Cisplatin is a highly emetogenic cancer chemotherapy agent, which is often used to induce nausea and emesis in animal models. The cytotoxic properties of cisplatin also cause adverse events that negatively impact on animal welfare preventing repeated administration of cisplatin. In this study, we assessed whether a low (subclinical) dose of cisplatin could be utilized as a model of nausea and emesis in the dog while decreasing the severity of adverse events to allow repeated administration. The emetic, nausea-like behavior and potential biomarker response to both the clinical dose (70 mg/m2) and low dose (15 mg/m2) of cisplatin was assessed. Plasma creatinine concentrations and granulocyte counts were used to assess adverse effects on the kidneys and bone marrow, respectively. Nausea-like behavior and emesis was induced by both doses of cisplatin, but the latency to onset was greater in the low-dose group. No significant change in plasma creatinine was detected for either dose groups. Granulocytes were significantly reduced compared with baseline (P = 0.000) following the clinical, but not the low-dose cisplatin group. Tolerability of repeated administration was assessed with 4 administrations of an 18 mg/m2 dose cisplatin. Plasma creatinine did not change significantly. Cumulative effects on the granulocytes occurred, they were significantly decreased (P = 0.03) from baseline at 3 weeks following cisplatin for the 4th administration only. Our results suggest that subclinical doses (15 and 18 mg/m2) of cisplatin induce nausea-like behavior and emesis but have reduced adverse effects compared with the clinical dose allowing for repeated administration in crossover studies.
Product Description
Mannitol, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis