  • Search Results
  • Photo-protective effect of americanin B against ultraviolet B-induced damage in cultured human keratinocytes.
Photo-protective effect of americanin B against ultraviolet B-induced damage in cultured human keratinocytes.
Environmental toxicology and pharmacology (2014-12-03)
Jian Zheng, Mei Jing Piao, Ki Cheon Kim, Cheng Wen Yao, Ji Won Cha, Jennifer H Shin, Suk Jae Yoo, Jin Won Hyun
ABSTRACT
Excessive ultraviolet (UV) radiation, a constituent of sunlight, can induce multiple types of skin damage. We recently demonstrated that americanin B, a lignin compound, protected cells against hydrogen peroxide (H2O2)-induced damage by exerting antioxidant effects and inhibiting apoptosis. In this study, we investigated the ability of americanin B to protect against cell injury induced by UVB (280-320nm), the most harmful UV wavelengths, in human HaCaT keratinocytes. Americanin B absorbed UVB, eliminated UVB-induced intracellular reactive oxygen species (ROS), and decreased the extent of UVB-induced oxidative modification of lipids, proteins, and DNA. In addition, americanin B inhibited UVB-induced apoptosis, as indicated by reductions in apoptotic body formation and DNA fragmentation. Furthermore, americanin B reversed the depolarization of the mitochondrial membrane induced by UVB exposure. These protective activities were associated with down-regulation of apoptosis-promoting proteins, Bax, caspase-9, and caspase-3 and up-regulation of an apoptosis inhibitor, Bcl-2. These results suggest that americanin B can protect human keratinocytes against UVB-induced cell damage.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
2′,7′-Dichlorofluorescein, ~90% (TLC), crystalline
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2′,7′-Dichlorofluorescein, ACS reagent
Sigma-Aldrich
JC-1, solid
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
2′,7′-Dichlorofluorescein, BioReagent, suitable for fluorescence, ≥90% (T)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Epigallocatechin gallate, primary reference standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)