  • Search Results
  • C-terminal domain small phosphatase 1 and MAP kinase reciprocally control REST stability and neuronal differentiation.
C-terminal domain small phosphatase 1 and MAP kinase reciprocally control REST stability and neuronal differentiation.
Proceedings of the National Academy of Sciences of the United States of America (2014-09-10)
Edmund Nesti, Glen M Corson, Maxwell McCleskey, Jon A Oyer, Gail Mandel
ABSTRACT
The repressor element 1 (RE1) silencing transcription factor (REST) in stem cells represses hundreds of genes essential to neuronal function. During neurogenesis, REST is degraded in neural progenitors to promote subsequent elaboration of a mature neuronal phenotype. Prior studies indicate that part of the degradation mechanism involves phosphorylation of two sites in the C terminus of REST that require activity of beta-transducin repeat containing E3 ubiquitin protein ligase, βTrCP. We identify a proline-directed phosphorylation motif, at serines 861/864 upstream of these sites, which is a substrate for the peptidylprolyl cis/trans isomerase, Pin1, as well as the ERK1/2 kinases. Mutation at S861/864 stabilizes REST, as does inhibition of Pin1 activity. Interestingly, we find that C-terminal domain small phosphatase 1 (CTDSP1), which is recruited by REST to neuronal genes, is present in REST immunocomplexes, dephosphorylates S861/864, and stabilizes REST. Expression of a REST peptide containing S861/864 in neural progenitors inhibits terminal neuronal differentiation. Together with previous work indicating that both REST and CTDSP1 are expressed to high levels in stem cells and down-regulated during neurogenesis, our results suggest that CTDSP1 activity stabilizes REST in stem cells and that ERK-dependent phosphorylation combined with Pin1 activity promotes REST degradation in neural progenitors.
Product Description
SAFC
L-Glutamine
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, ascites fluid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
Anti-MAP2 antibody, Mouse Monoclonal, clone HM-2, purified from hybridoma cell culture
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anti-MAP2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, ~2 mg/mL, clone AP-20, purified from hybridoma cell culture
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, clone AP-20, ascites fluid
Supelco
Acrylamide, analytical standard
Supelco
Acrylamide, certified reference material, TraceCERT®